Alcoholic Clarifier - Optik Type 2, Avantik
Supplier: Avantik
RS4367-B
76270-182CS 1196.47 USD
76270-182
Alcoholic Clarifier - Optik Type 2, Avantik
Stains Microscopy and staining reagents
Optik Type 2 Alcoholic Clarifier is designed to be used with Type 2 Hematoxylin to produce cell transparency and remove any non-specific staining. Slightly milder clarifier than the Optik Aqueous Clarifier.