Alcohol reagent ≥80% ACS

Supplier: Avantik

RS4078
76270-160CS 117.92 USD
76270-160
Alcohol reagent ≥80% ACS
Alcohol

A.C.S. grade. Meets and exceeds histological grade.


Avantik Alcohol is made from A.C.S. Grade Reagent Alcohol. The lower alcohol content varieties are made with A.C.S. Grade 100% Alcohol. The highest quality for all histology processing and staining procedures.

MDL Number: MFCD00003568
CAS Number: 64-17-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR