Alcohol reagent ≥100% ACS

Supplier: Avantik

Synonyms: Reagent alcohol

RS4029
76270-152CS 110.89 USD
76270-152
Alcohol reagent ≥100% ACS
Alcohol

A.C.S. grade. Meets and
exceeds histological grade.


Avantik Alcohol is made from A.C.S. Grade Reagent Alcohol. The lower alcohol content varieties are made with A.C.S. Grade 100% Alcohol. The highest quality for all histology processing and staining procedures.

MDL Number: MFCD00003568
CAS Number: 64-17-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR