Cole-Parmer® WD-200 Series Wax Dispensers, Antylia Scientific
Supplier: Argos Technologies
85230-08
76269-102EA 7246.66 USD
76269-102
Cole-Parmer® WD-200 Series Wax Dispensers, Antylia Scientific
Paraffin Wax Trimmers and Dispensers
Prevent wax blockage with heated tap.
  • Accurate temperature control with digital display
  • Constructed of stainless steel with antimicrobial coating to inhibit bacteria growth
  • Enhanced safety with high temperature cut-off at 120 °C
  • Fully insulated tank prevents heat loss and ensures the outer surface stays cool
This paraffin wax dispenser provides ultrafast heating for on-demand molten wax, ideal for histology and pathology applications. The heated tap prevents blockage providing a smooth even flow of wax every time. The nondrip lever has three settings - off, on-demand pull dispense, and continuous flow. The 0.5 mm mesh filter screen prevents coarse particles from entering the tap.
Temperature can be set from ambient to 70 °C using digital temperature control panel. The on/off temperature control keeps accuracy to ±1 °C. The 7.5 L capacity tank can melt up to 13.2 lbs. (6 kg) of pelletized wax at a time. When set at 65 °C, the unit will take approximately 90 minutes to fully melt 11 lbs. (5 kg) of pelletized wax.
Certifications: CE approved.
Delivery information: Supplied with power cord.