Tip Polisher, FT-700, American Hakko Products
Supplier: AMERICAN HAKKO PRODUCTS INC TE
FT700-05
76268-362EA 532.25 USD
76268-362
Tip Polisher, FT-700, American Hakko Products
Polishers and Tumblers
FT-700 Tip polisher helps increase the life of your tip with the action of two spinning polishing brushes.
  • Fast and easy way to keep your soldering iron tips clean and free from oxidation
  • Two spinning polishing brushes (brushes will not damage tip plating)
  • Lead-free chemical paste for immediate tip tinning
It is an expeditious and efficient way to boost tip life and keep free from oxidation. The tip polish, which has been specially formulated to boost the life of the tip.
Delivery information: Supplied with the tip polish and chemical paste.