G2R PCB Power Relay, Newark/Element
Supplier: NEWARK/ELEMENT 14 TE
G2R-24-DC12
76268-270EA 24.95 USD
76268-270
G2R PCB Power Relay, Newark/Element
Power Supplies
General purpose power Relays of single-pole10 A and double-pole 5 A.
Safety-oriented design with dielectric strength of 5,000 V between coil and contacts, and surge resistance of 10000 V. AC and DC types are both available for operational coils