Danger High Voltage Sign, Brady
Supplier: Brady Worldwide
84876
76268-222EA 25.86 USD
76268-222
Danger High Voltage Sign, Brady
Signs
This is an indoor/outdoor high performance pressure sensitive safety sign, and is surface printed polyester with a polyester overlaminate.
  • Weathers up to 8 years outdoors, even longer indoors, and withstands wide temperature variations
  • Permanent, pressure sensitive adhesive forms a long-lasting bond to even the most difficult surfaces and shapes
  • Cold temperature adhesive allows the signage to be applied at −18 °C
Flexible material conforms to most surfaces while protected graphics hold up to harsh environments, abrasion, and repeated spills and cleanings.