Shoe Cleaner, Ultra-Clean Products
Supplier: Ultra-Clean Products
1600SA-120V
76268-110EA 13699.82 USD
76268-110
Shoe Cleaner, Ultra-Clean Products
Vacuum Cleaners
Fully automatic shoe cleaner for use with external vacuum systems.
  • Simple to operate
  • Rugged design gives long life
  • Large footprint for stability
  • Speed control, brush motor
Five brushes for complete contamination removal.
Vacuum Exhaust - 1.50 I.D. x 1.75 O.D.
Size of main unit - 18"W x 27"L x 10.5"H
Overall height with handle - 40"
Weight - 50 LBS
Shipping Weight - 58 LBS
Voltage - 110-120V AC