Hot Air tool, Thermaltronics USA
Supplier: THERMALTRONICS USA INC TE
TMT-HA300-1
76267-944EA 654.15 USD
76267-944
Hot Air tool, Thermaltronics USA
Heaters Hot Air Dryers
The TMT-HA300 hot air tool uses a large diaphragm pump and can be used for surface mount component removal and reflow on components such as SOIC, CHIP, QFP, PLCC and others.
  • Digital display shows temperature and status
  • Airflow meter provides visual feedback of air flow
  • Large selection of high quality nozzles
  • Adjustable, easy to use temperature and air control knobs
Airflow (Max): 23 l/min
Pump: Diaphragm