Safety Cutter, 00200, Slice
Supplier: SLICE, INC.
00200
76267-908EA 15.05 USD
76267-908
Safety Cutter, 00200, Slice
Cutters
The 00200 Safety cutter is one of the first tools designed and remains one of the most popular. Initially conceived for household use, the safety cutter has found a home in industrial settings worldwide.
  • Blade type: micro-ceramic
  • Rubberized non-slip finish
  • Ambidextrous design, reduces injuries, lowers costs
  • Finger-friendly® edge stays sharp 11x longer
  • Clean-room friendly, non-sparking, non-conductive blade
  • Chemically inert blade never rusts
  • Built-in magnet and key-ring hole
The micro-ceramic blade cuts thin materials effectively while safeguarding fingers. Use the Safety Cutter's lanyard hole and built-in magnet to keep this unique tool close at hand. The product weighs 8.4g.