Protektive Pak® Small Component Shipper, Desco
Supplier: Protektive Pak
037000
76267-838EA 7.4 USD
76267-838
Protektive Pak® Small Component Shipper, Desco
Mailers and Shippers
Small Component Shipper for ESD sensitive items. Provides physical protection and reduces stress from physical shocks.
  • Static dissipative foam in Protektive Pak® box
  • Static dissipative surface resistance of 1×10⁶ to <1×10⁹ ohms per ANSI/ESD STM11.11
  • Buried shielding layer, minimizes sloughing and rub-off contamination
  • Made from 100% recycled material and is 100% recyclable
Pink static dissipative foam (1/2" thick) laminated to top, and conductive crosslink black polyethylene foam (1/4" thick) on bottom.
The Small Component Shipper provides an efficient means of shipping or storing small ESD sensitive items and offers ESD shielding with lid closed. “Faraday Cage” effect restricts electrostatic charges to exterior (All foam dimensions are ±1/16"). For physical protection, the foam cushioning reduces stress from physical shock. No assembly required.