Socket Adapter, Sid Tool Co
Supplier: SID TOOL CO INC TE
43966944
76267-782EA 105.11 USD
76267-782
Socket Adapter, Sid Tool Co
Tool Sets
Performance Series Socket Reducers make smaller square drive sockets and accessories compatible with 3/8" or 1/2" square drive drills or accessories.
  • Built for maximum durability and long life with impact drivers
  • Black oxide finish resists corrosion and increases life
Manufactured with heavy-duty, cold-forged, high-grade steel to withstand high torque output.