VinylStat FM2 ESD Anti-Fatigue Floor Matting, Transforming Technologies
Supplier: Transforming Technologies
FM22436GY
76267-706EA 114.76 USD
76267-706
VinylStat FM2 ESD Anti-Fatigue Floor Matting, Transforming Technologies
Floor Mats Traction and Anti-Fatigue Floor Mats
This reduces worker discomfort by providing a soft surface to stand on. Constructed with a expanded PVC that results in a cushioning spongy surface and provides a path-to-ground for electrostatic charges.
  • Provides a path-to-ground for ESD footwear
  • High grade thick closed cell PVC sponge for comfort and durability
Worker fatigue has been shown to lower productivity and morale which has a large impact on profitability. Damage due to static can have a similar drain on the bottom line, these problems can be solved.
Certifications: Meets ANSI ESD-S20.20 and ESD 4.1.