Jackson Safety® V50 Calico Safety Eyewear, Kimberly Clark
Supplier: Ansell Healthcare
25672
76267-642CS 329.25 USD
76267-642
Jackson Safety® V50 Calico Safety Eyewear, Kimberly Clark
Safety Glasses
Safety eyewear gives a lightweight protective solution that offers excellent wrap around and anti-fog protection in a stylish and comfortable design. The eyewear comes with temples that can be interchanged with a head strap for a closer fit.
  • Ideal for environments where there is a risk of impact or airborne debris
Safety glasses have smart features like a cushioned brow bar for additional comfort and ventilation, lenses with 99.9% UV protection to reduce eye strain and prevent eye damage like cataracts, retinal damage, and other conditions that can cause temporary vision loss, and a fog shield.
Wrap around design and impact resistant lenses make these safety goggles ideal for use in dirty manufacturing, transportation, metal manufacturing, as motorcycle goggles and many others.
Certifications: Meets EN166 1FT, CSA and ANSI Z87.1+ standards. EN166 7.3.1 resistance to surface damage. EN166 N anti fog coating and fog shield.