Moving Equipment Can Cause Severe Injury Keep Away Labels, Brady
Supplier: Brady Worldwide
96159
76267-570PK 48.85 USD
76267-570
Moving Equipment Can Cause Severe Injury Keep Away Labels, Brady
Signs
It is an indoor/outdoor high performance pressure sensitive safety sign, and is surface printed polyester with a polyester overlaminate.
  • Permanent, pressure sensitive adhesive
  • Forms a long lasting bond to even the most difficult surfaces and shapes
  • Abrasion and chemical resistant
Flexible material conforms to most surfaces while protected graphics hold up to harsh environments, abrasion, and repeated spills and cleanings.
Weathers up to 8 years outdoors, even longer indoors, and withstands wide temperature variations (up to 230 °F).
Particularly suited for outdoor applications, and in harsh environments where excellent abrasion and chemical resistance is required.