Multi Purpose Kit, Steinel America
Supplier: STEINEL AMERICA INC. TE
110051544
76267-564EA 969.36 USD
76267-564
Multi Purpose Kit, Steinel America
Heaters Hot Air Dryers
It provides heat gun torche.
Delivery information: Multi purpose kit includes: Reflector nozzle 9 mm; reflector nozzle 14 mm; reflector nozzle 39 mm; reduction nozzle 9 mm; welding nozzle; plastic welding rod assortment; plastic welding booklet; catalog; heavy duty case and HL 1920 E heat tool.