Xcelite® Screwdrivers for Slotted Screws, Apex
Supplier: Cooper Tools
R144N R184N R3322N R3323N R5166N
76241-478EA 47.39 USD
76241-478 76241-480 76242-334 76242-742 76250-634 76267-460
Xcelite® Screwdrivers for Slotted Screws, Apex
Screwdrivers
All blades are made of AISI 6150 chrome vanadium steel, plated for protection.
  • Available in regular round or pocket clip style round blade