Cole-Parmer® MP-250 Series Programmable Digital Melting Point Apparatus, Antylia Scientific
Supplier: Antyila Scientific
03010-41
76266-224EA 3685.13 USD
76266-224
Cole-Parmer® MP-250 Series Programmable Digital Melting Point Apparatus, Antylia Scientific
Melting Point Apparatus
Rotating viewing head ensures each user can analyze samples clearly.
  • Set ramp rate between 0.2° C/min to 10.0 °C in 0.1 °C increment
  • RS232 interface and USB ports allow for easy data transfer
  • Internal batch memory capacity of 1000 melts
  • View up to three samples simultaneously for greater efficiency
  • Height adjustable extension arm ensures proper viewing height for each user
  • Integral light and 8× magnifying lens enhance sample observation
  • Audible beeps indicate when temperature is stable and ready for sample
  • Extension arm folds away for easy storage
This versatile immersion circulator can be used in several ways. Use as a stand-alone water bath temperature controller, a pump for recirculating chilled liquid from an external chiller, or a pump for providing temperature control to an external device. Immersion circulator accurately heats up to 200 °C and safely controls the temperature of liquid in the bath while maintaining excellent temperature stability. A bidirectional RS232 interface lets you control bath remotely and download data to a computer for further analysis.
A digital display, overtemperature indicator light, pressure pump, and low-liquid level cutoff make the circulator easy and safe to use. Reach below ambient temperatures by using the cooling coil 12122-87 connected to an optional external chiller.
Delivery information: Supplied with 100 capillary tubes, lens hood, power supply, and power cord sets with US, UK, and EU plugs.