SafeCup™ III MultiDrug 14 Panel, Germaine® Laboratories
Supplier: Germaine Laboratories
44418
76266-174CS 591.62 USD
76266-174
SafeCup™ III MultiDrug 14 Panel, Germaine® Laboratories
Drug Tests
SafeCup™ lll Round Cups are an all-in-one (collection/testing/transport) qualitative test for the detection of drugs of abuse in urine.
  • Tests adhere to SAMSHA cutoff levels
  • Leak-proof cap, temperature strip, & security seal included
All-in-One Cup tests for AMP, BAR, BZO, BUP, COC, MTD, MDMA, MET, OPI, OXY, PCP, TCA, THC, and PPX. CLIA Waived. Includes 25 collection cups, security seals, gloves, and instructions.