UA Liquid Controls Plus, Germaine Laboratories
Supplier: Germaine Laboratories
51116
76266-172BX 216.01 USD
76266-172
UA Liquid Controls Plus, Germaine Laboratories
Controls and Calibration Verification Materials
UA liquid controls plus is an assayed urine control intended for use in validating the precision of visual and analyzer reading of urinalysis.
Use for one or more of the following analytes: Ascorbic Acid, Glucose, Bilirubin, Ketone (Acetoacetic Acid), Specific Gravity, Blood, pH, Protein, Urobilinogen, Nitrite, Leukocytes, and Human Chorionic Gonadotropin.