AimTab™ Reducing Substances Tablets, Germaine Laboratories
Supplier: Germaine Laboratories
13320
76266-170PK 42.7 USD
76266-170
AimTab™ Reducing Substances Tablets, Germaine Laboratories
Clinical Diagnostic Supplies Clinical Reagents and Kits
AimTab™ Reducing Substances Tablets are for the semi-quantitative determination for the amount of reducing substances (generally glucose) in urine, stool, wine, and other specimens.