Assortment Kits - Adapters and Stoppers, Ace Glass Incorporated
Supplier: Ace Glass
5701-01 5701-02 5701-03 5701-13 5701-14 5701-21
76266-064EA 564.36 USD
76266-064 76266-066 76266-068 76266-072 76266-074 76266-076
Assortment Kits - Adapters and Stoppers, Ace Glass Incorporated
Adapters Adapters, Joints with Tube Connectors
Assortment kits ensure the right adapter or stopper is on hand when it is needed most. Choose from reducing and enlarging standard taper glass joint adapters, Ace-Thred probe adapters, and hollow glass stoppers.