Amylo-Glo RTD Amyloid Plaque Stain Reagent, Biosensis
Supplier: Biosensis
TR-300-AG
76264-656EA 459 USD
76264-656
Amylo-Glo RTD Amyloid Plaque Stain Reagent, Biosensis
Stains Histology Stains
Amylo-Glo® RTD™ 'Ready to Dilute' Staining reagent is designed to stain amyloid plaques in tissue sections.
  • For Amyloid plaques - Both intraneuronal and vascular
  • Very bright blue UV excitable stain under physiological conditions
  • Unique excitation/emission profile and mild staining
  • Combination for multiple immunofluorescent labeling studies
  • Storage Up to 6 months after date of receipt at 4 °C protected from light
This novel marker has several advantages over other conventional markers such as Thioflavin S and Congo Red because of its unique chemical and spectral properties. (L. Schmued et al. (2012) J.Neuroscience Methods 209:120– 126). Using Amylo-Glo® results in a very bright blue UV excitable stain under physiological conditions that will not bleed through when illuminated with other filters. Its brightness makes it ideal for low magnification quantification studies, while its unique excitation/emission profile and mild staining conditions makes it ideal for combination for multiple immunofluorescent labeling studies.
Amylo-Glo® RTD™ is compatible with fresh, frozen, and formalin-fixed immunohistochemistry or cytochemistry, and it is particularly good for confocal and multiple labeling because of its high fluorescent intensity and high resistance to photo-bleaching. Moreover because Amylo-Glo® fluoresces in the UV channel, double and triple labeling experiments can be performed very easily (see protocol). Reagent detects amyloid plaques, both intraneuronal and vascular.