Waterproof Cut Resistant Gloves, Showa
Supplier: Showa
S-TEX377S-06 S-TEX377M-07 S-TEX377L-08 S-TEX377XL-09 S-TEX377XXL-10
76264-560CS 1980.04 USD
76264-560 76264-562 76264-564 76264-566 76264-568
Waterproof Cut Resistant Gloves, Showa
Gloves
A flexible and robust fully coated nitrile glove to protect against water, grease, oil and abrasion with high cut resistance. The additional nitrile foam palm coating provides outstanding grip in wet and oily conditions.
  • Cut resistance level 4
  • Full coating of flat nitrile with nitrile foam palm
  • Seamless knitted liner prevents irritation
  • Lightweight, 13 gauge liner allows excellent hand dexterity
  • Allows movement of hand as the hand flexes
  • It has a liner material of HAGANE Steel Coil®
Manufactured using patented hand former design for a comfortable fit and to minimize hand fatigue.
Cuff is longer to extend protection. The elasticated knitted wrist helps to keep the glove in place.
Certifications: CE Cat II marked. EN 388:2016-4441D.