Flexible Neoprene Gauntlets with Rough Textured Finish, Showa
Supplier: Showa
3415-09 3415-10 3415-11
76264-532CS 350.78 USD
76264-532 76264-534 76264-536 76264-538
Flexible Neoprene Gauntlets with Rough Textured Finish, Showa
Gloves
The pinnacle of protection, comfort and dexterity. Gauntlet glove creates a defensive barrier against a range of chemicals, including acids, caustics, solvents, grease and oil.
  • Flexible neoprene coating
  • Fully coated
  • Rough textured finish on palm
  • Level 3 abrasion resistance
The seamless liner with 15 gauge prevents irritation.
Certifications: EN 388: 2003-3031. EN 374-2 - 2003. EN 374:2003.