WHEATON® Microwave Extraction Tube, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
45047-30200
76264-508CS 78.58 USD
76264-508
WHEATON® Microwave Extraction Tube, DWK Life Sciences
Extractors Soxhlet and Soxhlet-Type Extractors
Manufactured from 33 expansion borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
  • Round bottom
  • Unmarked
Certifications: Conforming to USP Type I and ASTM E438, Type I, Class A requirements.