VWR® Blue Twist-Seal Bags
Supplier: VWR International
76264-506
76264-506CS 579.89 USD
76264-506
VWR® Blue Twist-Seal Bags
Bags Sample Bags
These blue twist-seal printed bags with safety tab and 2 round wires are made of virgin PE and have been specifically designed for the food industry.
  • Blue color tear-off for increased visibility
  • Distinct color to avoid possible foreign matter contamination
  • Visual recognition of the product, but can also be used to segregate within a factory environment
  • Optimized opening area to facilitate insertion of sample
  • Lot number traceability
  • All sterile sampling bags are also free from RNase, DNase and pyrogens
The bags accommodate solid, semi-solid and liquid compounds.
Certifications: Food contact approved.