Fully Coated Latex, Water Repellent and Thermal Gloves, Showa
Supplier: Showa
406XL-09 406XXL-10 406M-07 406L-08
76264-498CS 1694.65 USD
76264-498 76264-500 76264-570 76264-572
Fully Coated Latex, Water Repellent and Thermal Gloves, Showa
Gloves
A durable, water repellent, insulated grip glove that’s designed to protect down to –30 °C for short or intermittent contact.
  • Thermal liner
  • Full foam latex coating with second latex coating on palm
  • Orange color enhances safety in low light conditions
  • Rough textured finish to palm coating
  • Keeps hand warm and dry
Aerated material reduces heat loss via conduction, and eliminates convection by trapping warm air inside the glove. All while the engineered liner and foam latex dissipate sweat and moisture to prevent hands from getting cold.
Coating technology enhances a high level of flexibility and reduces fatigue. Glove maintains a soft, comfortable fit, while the seamless liner prevents irritation.
Certifications: CE Cat. II marked. EN 388: 2003-2131. EN 511: 2006.