Eco Best® Biodegradable, Chemical Resistant, Hi-Vis Gloves, Showa
Supplier: Avantor
76264-488CS 371.9 USD
76264-488 76264-492 76264-494 76264-496 76264-520
Eco Best® Biodegradable, Chemical Resistant, Hi-Vis Gloves, Showa
Gloves
Complete biodegradable nitrile disposable glove with Eco Best Technology® in a hi-vis orange color protects against oils, hydrocarbons, grease and many chemicals.
  • Fits like second skin
  • Fluorescent orange color increases visibility
  • Bisque finish on palm and fingers provides a long-lasting grip
  • Impermeable, for working in damp or greasy environments
  • Rolled cuff prevents dirt from entering the glove
  • Chlorinated for ease of donning and doffing
Certifications: CE Cat. III marked. EN388:2016, 1001X. EN374-1:2016/Type A. EN374-5:2016. Directive 2002/72 EC.