Synthware Single Volume Pipettes, Kemtech America
Supplier: Kemtech America
P191001 P191002 P191025 P191050
76264-332EA 14.42 USD
76264-332 76264-334 76264-336 76264-338 76264-340 76264-342 76264-344
Synthware Single Volume Pipettes, Kemtech America
Pipettes Micropipettes
These single volume pipettes are certified class A.
  • High quality borosilicate glass
Synthware single volume pipets are perfect for transferring small amounts of liquid.
Certifications: Meets ASTM Specification E438, Type 1, Class A, and 'Standard Specification for Glasses in Laboratory Apparatus'.