Stainless Steel Micro Spatula, United Scientific Supplies
Supplier: United Scientific Supplies
SSFT08-VWR
76263-356EA 4.15 USD
76263-356
Stainless Steel Micro Spatula, United Scientific Supplies
Spatulas
Stainless steel micro spatula features one end flat and one end pointed.
  • Stainless steel construction
  • One pointed end
This micro spatula is perfect for mixing and dispensing small volume of powders or solutions.