Funnels with Short Stems, United Scientific Supplies
Supplier: United Scientific Supplies
FHD165 FHD185 FHD125
76263-216EA 31.07 USD
76263-216 76263-218 76263-220
Funnels with Short Stems, United Scientific Supplies
Funnels Powder Funnels
Funnels feature short, wide stems, and are useful for adding powder chemical to reaction vessels.
  • Wide stems
  • Short stems to avoid clogging
The 35 ml and 300 ml sizes are made of high density polyethylene, and the 100 ml size is made of polypropylene.