UniStore™ Reagent Bottles, Wide Mouth, PP
Supplier: United Scientific Supplies
33311 33306 33307 33308 33309 33310 33311-BULK 33306-BULK 33307-BULK 33308-BULK 33309-BULK 33310-BULK
76263-108PK 11.99 USD
76263-108 76263-110 76263-112 76263-114 76263-116 76263-118 470340-138 470338-890 470339-158 470339-242 470340-026 470339-928
UniStore™ Reagent Bottles, Wide Mouth, PP
Bottles Plastic Bottles
Bottles are for general laboratory use and for storing, shipping, and packing liquids.
  • Translucent
  • Rigid
  • Autoclavable
UniStore™ wide mouth PP reagent bottles are manufactured from USP Class VI resin that is FDA-21CFR compliant. Excellent chemical resistance to most acids, bases, and alcohols.
Ordering information: Polypropylene caps are included.