UniStore™ Reagent Bottles, Wide Mouth, HDPE
Supplier: United Scientific Supplies
33406 33407 33408 33409 33410 33406-BULK 33407-BULK 33408-BULK 33409-BULK 33410-BULK 33412 33414 33441
76263-098PK 22.88 USD
76263-098 76263-100 76263-102 76263-104 76263-106 470339-008 470339-472 470339-130 470340-098 470339-570 470349-772 470349-760 470349-784
UniStore™ Reagent Bottles, Wide Mouth, HDPE
Bottles Plastic Bottles
Bottles are for general laboratory use and for storing, shipping, and packing liquids.
  • Translucent
  • Pliable
UniStore™ wide mouth HDPE reagent bottles are manufactured from USP Class VI resin that is FDA-21CFR compliant. Excellent chemical resistance to most acids, bases, and alcohols.
Ordering information: Polypropylene caps are included.