Beaker Tongs, United Scientific Supplies
Supplier: United Scientific Supplies
BTSP09-VWR
76263-096EA 9.38 USD
76263-096
Beaker Tongs, United Scientific Supplies
Tongs
These long-lasting beaker tongs are made of nickel-plated steel.
  • Maximum jaw spread: 17 cm
  • Vinyl coated sleeves
  • Overall length is 25.7 cm
Beaker tongs feature strong, zinc-plated steel construction with flat riveted joints and vinyl coated sleeves that open to 7". Tongs can handle beakers ranging from 50 to 1500 ml capacity. Overall length is 9.75".