Beakers with Handles, United Scientific Supplies
Supplier: United Scientific Supplies
81121 81122 81123 81124 81105-VWR PJSET5
76263-072PK 50.53 USD
76263-072 76263-074 76263-076 76263-078 76263-080 470338-874
Beakers with Handles, United Scientific Supplies
Beakers
All sizes have handles for convenient lifting.
  • Autoclavable
  • Good chemical resistance
  • All sizes have handles for convenient lifting
Short form beakers with raised graduations, molded in polypropylene.