Anti-PIK3CB Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
600-401-GT8
76262-946EA 551.64 USD
76262-946
Anti-PIK3CB Rabbit Polyclonal Antibody
Antibodies
PIK3CB is a catalytic subunit of phosphoinositide-3-kinase beta. PIK3CB catalyzes the production of phosphatidylinositol-3,4,5-triphosphate by phosphorylating phosphatidylinositol (PI), phosphatidylinositol-4-phosphate (PIP) and phosphatidylinositol-4,5-bisphosphate (PIP2). Growth factors and hormones trigger this phosphorylation event, which in turn coordinates cell growth, cell cycle entry, cell migration and cell survival. PTEN reverses this process. PI3K signaling pathway is constitutively activated in human cancers that have loss of function of PTEN. Furthermore, PI3KCB down regulation can suppress cell growth in malignant gliomas. Anti-PIK3C beta Antibody is suitable for researcher in Cancer, Immunology and Nuclear Signaling research.

Anti-PIK3C beta Antibody has been tested for use in ELISA and western blotting.  Specific conditions for reactivity should be optimized by the end user.  Expect a band approximately 122.7 kDa in size corresponding to PI3K beta by western blotting in the appropriate cell lysate or extract.

Type: Primary
Antigen: PIK3CB
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR