Anti-MAP2K1 Rabbit Polyclonal Antibody (DL680)

Supplier: Rockland Immunochemical
600-444-GN7S
76262-886EA 150.22 USD
76262-886
Anti-MAP2K1 Rabbit Polyclonal Antibody (DL680)
Antibodies
MEK1 antibodies detect the MEK1 isoform. Mitogen-activated protein kinase kinase 1, also known as MKK or MEK, is an integral component of the MAP kinase cascade that regulates cell growth and differentiation. This pathway also plays a key role in synaptic plasticity in the brain. Activated MEK 1 acts as a dual specificity kinase phosphorylating both a threonine and a tyrosine residue on MAP kinase. The MEK1 antibody is ideal for investigators involved in Neuroscience, Cell Signaling and Cancer Research.

Anti-MEK 1 (RABBIT) antibody is suitable for use in ELISA and Western Blotting. Specific conditions of reactivity should be optimized by the end user. Expect a band of approximately 43.5 kDa.

Type: Primary
Antigen: MAP2K1
Clonality: Polyclonal
Clone:
Conjugation: DL680
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR