Anti-MAP2K2 Rabbit Polyclonal Antibody (BAC)

Supplier: Rockland Immunochemical
600-406-GQ2S
76262-876EA 150.22 USD
76262-876
Anti-MAP2K2 Rabbit Polyclonal Antibody (BAC)
Antibodies
MEK2 antibodies detect the MEK2 isoform. Mitogen-activated protein kinase kinase 2, also known as MEK2 or MKK2, is an integral component of the MAP kinase cascade that regulates cell growth and differentiation. This pathway also plays a key role in synaptic plasticity in the brain. Activated MEK 2 acts as a dual specificity kinase phosphorylating both a threonine and a tyrosine residue on MAP kinase. MEK1 and MEK2 are about 80% identical to each other, and nearly identical within the kinase domain. The MEK2 antibody is ideal for investigators involved in Neuroscience, Cell Signaling and Cancer Research.

Anti-MEK 2 (RABBIT) antibody is suitable for use in ELISA and Western Blotting. Specific conditions of reactivity should be optimized by the end user. Expect a band of approximately 44 kDa.

Type: Primary
Antigen: MAP2K2
Clonality: Polyclonal
Clone:
Conjugation: BAC
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR