Anti-MAPK3 Rabbit Polyclonal Antibody (BAC)

Supplier: Rockland Immunochemical
600-406-GP0S
76262-856EA 150.22 USD
76262-856
Anti-MAPK3 Rabbit Polyclonal Antibody (BAC)
Antibodies
ERK1 antibodies detect the ERK1 isoform. Mitogen activiated protein kinase 3, also known as MAPK3, ERK, or ERK1, is an integral component of the MAP kinase cascade that regulates cell growth and differentiation. ERK1 and ERK2 are activated by MEK1 and MEK2 in the B-raf signaling pathway resulting in its translocation to the nucleus where it phosphorylates nuclear targets. Human ERK1 and ERK2 are 84% identical in sequence and share common functionality in cells. Anti-ERK1 antibody is ideal for investigators involved in Neuroscience, Cell Signaling and Cancer Research.

Anti-ERK 1 (RABBIT) antibody is suitable for use in ELISA and Western Blotting. Specific conditions of reactivity should be optimized by the end user. Expect a band of approximately 43 kDa.

Type: Primary
Antigen: MAPK3
Clonality: Polyclonal
Clone:
Conjugation: BAC
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR