Anti-cas9 Rabbit Polyclonal Antibody (HRP)

Supplier: Rockland Immunochemical
600-403-GK0S
76262-816EA 151.49 USD
76262-816
Anti-cas9 Rabbit Polyclonal Antibody (HRP)
Antibodies
The Clustered Regularly Interspaced Short Palindromic Repeats (CRISPR) and CRISPR Associated (Cas) system was discovered in bacteria as a defense against foreign DNA, even being believed to act as a sort of prokaryotic immune system. The Cas9 protein is becoming a a useful tool in the field of genomic editing for its ability to induce site-directed double strand breaks in DNA. It can cleave almost any sequence complementary to the guide RNA and has recently been used for modifying the genome of human embryos for the first time.

Cas9 Peroxidase Conjugated Antibody has been tested for use in ELISA, Immunofluorescence, and western blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band ~158 kDa in size corresponding to Cas-9 protein by western blotting in the appropriate cell lysate or extract.

Type: Primary
Antigen: cas9
Clonality: Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR