Anti-COL6A1 Rabbit Polyclonal Antibody (FITC)

Supplier: Rockland Immunochemical
600-402-108
76262-776EA 575.67 USD
76262-776
Anti-COL6A1 Rabbit Polyclonal Antibody (FITC)
Antibodies
Anti-Collagen Type VI Fluorescein Conjugated Antibody is specific for collagen type VI protein. The collagens are a superfamily of proteins that play a role in maintaining the integrity of various tissues. Collagen VI is a major structural component of microfibrils. The basic structural unit of collagen VI is a heterotrimer of the alpha1(VI), alpha2(VI), and alpha3(VI) chains. The alpha2(VI) and alpha3(VI) chains are encoded by the COL6A2 and COL6A3 genes, respectively. The protein encoded by this gene is the alpha 1 subunit of type VI collagen (alpha1(VI) chain). Mutations in the genes that code for the collagen VI subunits result in the autosomal dominant disorder, Bethlem myopathy.

Anti-Collagen Type VI (RABBIT) Fluorescein Conjugated Antibody is suitable for western blot, immunoprecipitatioin, and immunohistochemistry. Researchers should determine optimal titers for applications that are not stated below.

Type: Primary
Antigen: COL6A1
Clonality: Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR