Anti-COL2A1 Rabbit Polyclonal Antibody (FITC)

Supplier: Rockland Immunochemical
600-402-104
76262-768EA 575.67 USD
76262-768
Anti-COL2A1 Rabbit Polyclonal Antibody (FITC)
Antibodies
Anti-Collagen Type II Fluorescein Conjugated Antibody detects Type-II collagen which is the basis for articular cartilage and hyaline cartilage. It makes up 50% of all protein in cartilage and 85-90% of collagen of articular cartilage. Type II collagen does form fibrils which allow cartilage to entrap the proteoglycan aggregate as well as provide tensile strength to the tissue. Anti-Collagen Type II antibody is ideal for investigators involved in Immunology, Neuroscience and Signal Transduction research.

Anti-Collagen Type II Fluorescein Conjugated Antibody is suitable for western blotting, IP, IHC and for ELISA. Researchers should determine optimal titers for applications that are not stated below.

Type: Primary
Antigen: COL2A1
Clonality: Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR