Anti-Ffar4 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
600-401-GW0S
76262-736EA 151.49 USD
76262-736
Anti-Ffar4 Rabbit Polyclonal Antibody
Antibodies
Anti-Ffar4 Antibody was designed, produced, and validated as part of the Joy Cappel Young Investigator Award (JCYIA). Free Fatty Acid 4 receptor (Ffa4 receptor or GPR120), a rhodopsin-like G protein coupled receptor (GPCR) subfamily member, is a receptor that senses specific fatty acids such as omega-3 fatty acid in fish oil or the endogenous signaling lipid, PHASA. Ffa4 receptor is enriched in lung, colon and adipose tissue but is also detected in many other tissues and cells. The activation of Ffar4 has multiple effects, including but not limited to inhibition of inflammation, improving insulin sensitivity and adipogenesis, and regulating hormone secretion from the gastro-intestinal system and pancreatic islets. Therefore, approaches that regulate FFA4 receptor activity could be developed as promising anti-diabetic and anti-inflammation drugs. GPR120 is the only fatty acid receptor that can sense lipids in adipose tissue, mature adipocytes, CD11c+ macrophages, and RAW264.7 cells making this receptor of potential importance in the prevention and treatment of metabolic and inflammatory diseases.

Anti-Ffar4 Antibody has been tested for use in ELISA and western blot.  Specific conditions for reactivity should be optimized by the end user. Expect a band ~40-48 kDa in size corresponding to Ffar4 by western blotting in the appropriate cell lysate or extract. Tested using positive controls Raw 264.7 WCL p/n W10-001-369 and NIH/3T3 WCL p/n W10-000-358.

Type: Primary
Antigen: Ffar4
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR