Anti-TJP1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
600-401-GU7S
76262-716EA 151.49 USD
76262-716
Anti-TJP1 Rabbit Polyclonal Antibody
Antibodies
ZO-1, also called TJP1, belongs to the MAGUK family. This gene encodes a protein located on a cytoplasmic membrane surface of intercellular tight junctions. The encoded protein may be involved in signal transduction at cell-cell junctions. The N-terminal may be involved in transducing a signal required for tight junction assembly, while the C-terminal may have specific properties of tight junctions. The alpha domain might be involved in stabilizing junctions. ZO-1 plays a role in the regulation of cell migration by targeting CDC42BPB to the leading edge of migrating cells. ZO1 may be associated the following disorders; celiac desease, congenital nephrotic syndrome finnish type, and macular degeneration. Anti-ZO-1 Antibody is useful for researchers interested in Apoptosis Research and Insulin Research.

Anti-ZO-1 Antibody has been tested in Western Blot, ELISA, Immuohistochemistsry, Immunofluorescence, and Flow Cytometry. Expect a band at ~245 and/or 195.5 kDa in western blot using appropriate lysates. Positive control whole cell lysates used A549 and PC3 @ 1µg/mL for WB; CACO2 and PC3 with PFA and MeOH @ 10µg/mL for IF. Positive control cells for FC were PC3 and positive control tissues for IHC was mouse adipose tissue.

Type: Primary
Antigen: TJP1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR