Anti-BCL3 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
600-401-GU4
76262-710EA 551.64 USD
76262-710
Anti-BCL3 Rabbit Polyclonal Antibody
Antibodies
BCL3 belongs to the ANKRD family. The protein contains seven ankyrin repeats, which are closely related to those found in I kappa B proteins. BCL3 functions as a transcriptional co-activator, regulator of transcriptional activation, promotor of transcription, and inhibitor of nuclear translocation of NF-kappa B. It is a proto-oncogene identified by its translocation into the immunoglobulin alpha-locus in some cases of B-cell leukemia. BCL3 contributes to the regulation of cell proliferation. Therefore, BCL3 may be associated with B-cell chronic lymphocytic leukemia, lymphoma, or other hematologic diseases. Anti-BCL3 Antibody is useful for researchers interested in Cancer Research, NF-Kappa B Pathway, and TNF Signaling Pathway.

Anti-BCL-3 Antibody has been tested in Western Blot, ELISA, and Immunofluorescence. Expect a band at 47.5 kDa in western blot using appropriate lysates. Positive control used recombinant BCL3 protein, HEK293T overexpressing BCL3 or U87 whole cell lysates at 1µg/mL in WB; and U87MG with PFA at 10µg/mL in IF. Although not tested, this antibody is likely functional in immunohistochemistry, flow cytometry, and immunoprecipitation.

Type: Primary
Antigen: BCL3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR