Anti-SLC7A11 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
600-401-GU3
76262-706EA 551.64 USD
76262-706
Anti-SLC7A11 Rabbit Polyclonal Antibody
Antibodies
xCT belongs to the amino acid-polyamine-organocation (APC) superfamily and L-type amino acid transporter (LAT) family. xCT gene encodes a member of a heteromeric, sodium-independent, anionic amino acid transport system that is highly specific for cysteine and glutamate. In this system, designated Xc(-), the anionic form of cysteine is transported in exchange for glutamate. Increased expression of this gene results in neuronal cell death. xCT may be associated with kaposi sarcoma, dyscalculia, cystinuria, spondylolsis, and anemia of prematurity. Anti-xCT Antibody is useful for researchers interested in Cancer Research, Glucose/Energy Metabolism Research, and Cell Surface Protein Interaction Research.

Anti-xCT Antibody has been tested in Western Blot, ELISA, Immunofluorescence and Flow cytometry. Expect a band at 55.4 kDa in western blot using appropriate lysates. Positive control used NIH-3T3, Hela, HCT-116, A549, or PC3 whole cell lysates at 0.1-1.0µg/mL for WB and PC3 with MeOH at 10µg/mL in IF. Although not tested, this antibody is likely functional in immunohistochemistry and immunoprecipitation.

Type: Primary
Antigen: SLC7A11
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR