Anti-SERPINA1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
600-401-147-0100
76262-656EA 551.64 USD
76262-656
Anti-SERPINA1 Rabbit Polyclonal Antibody
Antibodies
ALPHA-1-ANTI-TRYPSIN Antibody is specific for alpha-1-anti-trypsin. Alpha 1-Antitrypsin or α1-antitrypsin (A1AT) is a protease inhibitor belonging to the serpin superfamily and is generally known as serum trypsin inhibitor. Alpha-1-anti-trypsin protects tissues from enzymes of inflammatory cells. A1AT antibody is suitbale for immunology and cardiovascular researcher.

Anti-Human ALPHA-1-ANTI-TRYPSIN Antibody has been assayed against 1.0 µg of Human Alpha-1-anti-Trypsin in a standard ELISA using Peroxidase conjugated Affinity Purified Donkey anti-Rabbit IgG and TMB as a substrate for 30 minutes at room temperature.  A working dilution of 1:10,000 to 1:50,000 is suggested for this product.

Type: Primary
Antigen: SERPINA1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR