Oakton® EcoTestr™ Conductivity Waterproof Pocket Testers, Cole-Parmer
Supplier: Argos Technologies
35634-06 WD-35634-08
76258-454EA 161.93 USD
76258-454 76258-456
Oakton® EcoTestr™ Conductivity Waterproof Pocket Testers, Cole-Parmer
Electrochemistry Meters Conductivity Meters
Designed to be durable and easy to use.
  • View measurements clearly on large LCD
  • Rugged housing is waterproof, dustproof, and floats in water
  • Secure, attach a lanyard to prevent losing or dropping in the field
  • Auto shutoff extends battery life
  • Two years warranty
Leakproof cap with fill line functions as a sample/calibration and hydrating cup.
Certifications: CE marked, IP 67 rated housing.
Delivery information: Includes plastic storage case, lanyard, and four AAA batteries.