3M™ Cable Ties, 3M
Supplier: 3M
7000133454 7000031849
76244-050CS 34.41 USD
76244-050 76254-078
3M™ Cable Ties, 3M
Ties
3M™ Cable ties are molded from 6/6 nylon and are available in a variety of sizes and styles, including weather resistant black for outdoor use. They are used to secure wire bundles and harness components quickly. The flexible design allows the user to slip ties easily under and around cables and harnesses. These ties can be used with most standard tensioning/cutoff tools for fast production line fastening.